3-Chloro-1-methyl-1H-1,2,4-triazole

Supplier: Matrix Scientific

080973-500MG 080973-1G 080973-5G
102672-168EA 646.15 USD
102672-168 102672-170 102672-172
3-Chloro-1-methyl-1H-1,2,4-triazole
3-Chloro-1-methyl-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₃H₄ClN₃
MDL Number: MFCD22422027
CAS Number: 56616-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR