[2-(1H-Tetrazol-5-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

080968-500MG 080968-1G 080968-5G
102672-144EA 646.15 USD
102672-144 102672-146 102672-148
[2-(1H-Tetrazol-5-yl)ethyl]amine hydrochloride
[2-(1H-Tetrazol-5-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₃H₈ClN₅
MDL Number: MFCD09864345
CAS Number: 33841-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR