(1H-Tetrazol-5-ylmethyl)amine hydrochloride

Supplier: Matrix Scientific

080967-500MG 080967-1G 080967-5G
102672-138EA 516.92 USD
102672-138 102672-140 102672-142
(1H-Tetrazol-5-ylmethyl)amine hydrochloride
(1H-Tetrazol-5-ylmethyl)amine hydrochloride

Hazard: Irritant

Formula: C₂H₆ClN₅
MDL Number: MFCD11857773
CAS Number: 31602-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR