2,9-Diazaspiro[5.5]undecan-1-one hydrochloride

Supplier: Matrix Scientific

080966-500MG 080966-1G 080966-5G
102672-132EA 646.15 USD
102672-132 102672-134 102672-136
2,9-Diazaspiro[5.5]undecan-1-one hydrochloride
2,9-Diazaspiro[5.5]undecan-1-one hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O
MDL Number: MFCD08461145
CAS Number: 1187173-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR