6-Iodoquinazolin-4-amine

Supplier: Matrix Scientific

080963-500MG 080963-1G 080963-5G
102672-114EA 516.92 USD
102672-114 102672-116 102672-118
6-Iodoquinazolin-4-amine
6-Iodoquinazolin-4-amine

Hazard: Irritant

Formula: C₈H₆IN₃
MDL Number: MFCD03538049
CAS Number: 182880-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR