4,5,6,7-Tetrahydro-2H-indazol-3-ol

Supplier: Matrix Scientific

080962-500MG 080962-1G 080962-5G
102672-108EA 258.46 USD
102672-108 102672-110 102672-112
4,5,6,7-Tetrahydro-2H-indazol-3-ol
4,5,6,7-Tetrahydro-2H-indazol-3-ol

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD00462221
CAS Number: 402602-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR