2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
Supplier: Matrix Scientific
Synonyms:
2-Amino-4-hydroxy-5,6,7,8-tetrahydroquinazoline
080960-500MG
080960-1G
080960-5G
102672-096EA
387.69
USD
102672-096
102672-098
102672-100
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
2-Amino-5,6,7,8-tetrahydroquinazolin-4-ol
Hazard: Irritant
Formula:
C₈H₁₁N₃O MW: 165.2 g/mol |
MDL Number:
MFCD00665850 CAS Number: 33081-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...