1H-Benzimidazole-5-sulfonic acid

Supplier: Matrix Scientific

080958-500MG 080958-1G
102672-088EA 387.69 USD
102672-088 102672-090
1H-Benzimidazole-5-sulfonic acid
1H-Benzimidazole-5-sulfonic acid

Hazard: Irritant

Formula: C₇H₆N₂O₃S
MDL Number: MFCD06617940
CAS Number: 27503-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR