Methyl-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate

Supplier: Matrix Scientific

080956-500MG 080956-1G 080956-5G
102672-076EA 138.46 USD
102672-076 102672-078 102672-080
Methyl-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate
Methyl-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropanecarboxylate

Hazard: Irritant

Formula: C₉H₁₂Cl₂O₂
MDL Number: MFCD00044765
CAS Number: 59897-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR