3-(Chloromethyl)-1-methylpiperidine hydrochloride

Supplier: Matrix Scientific

080952-500MG 080952-1G 080952-5G
102672-060EA 516.92 USD
102672-060 102672-062 102672-064
3-(Chloromethyl)-1-methylpiperidine hydrochloride
3-(Chloromethyl)-1-methylpiperidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₅Cl₂N
MW: 184.11 g/mol
MDL Number: MFCD00012833
CAS Number: 66496-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR