3-(Chloromethyl)-4-methyl-4H-1,2,4-triazole hydrochloride

Supplier: Matrix Scientific

080951-500MG 080951-1G
102672-056EA 550.77 USD
102672-056 102672-058
3-(Chloromethyl)-4-methyl-4H-1,2,4-triazole hydrochloride
3-(Chloromethyl)-4-methyl-4H-1,2,4-triazole hydrochloride

Hazard: Irritant

Formula: C₄H₇Cl₂N₃
MDL Number: MFCD20483332
CAS Number: 135206-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR