(4-Methyl-4H-[1,2,4]triazol-3-yl)methanol

Supplier: Matrix Scientific

080950-500MG 080950-1G
102672-052EA 516.92 USD
102672-052 102672-054
(4-Methyl-4H-[1,2,4]triazol-3-yl)methanol
(4-Methyl-4H-[1,2,4]triazol-3-yl)methanol

Hazard: Irritant

Formula: C₄H₇N₃O
MW: 113.1200 g/mol
MDL Number: MFCD13190102
CAS Number: 59660-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR