[3-(Aminomethyl)phenyl]methanol
Supplier: Matrix Scientific
Synonyms:
3-(Aminomethyl)benzyl Alcohol
080948-500MG
080948-1G
080948-5G
102672-040EA
516.92
USD
102672-040
102672-042
102672-044
[3-(Aminomethyl)phenyl]methanol
[3-(Aminomethyl)phenyl]methanol
Hazard: Irritant
Formula:
C₈H₁₁NO |
MDL Number:
MFCD06212981 CAS Number: 34231-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...