(5-Mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methanol

Supplier: Matrix Scientific

080947-500MG 080947-1G
102672-036EA 387.69 USD
102672-036 102672-038
(5-Mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methanol
(5-Mercapto-4-methyl-4H-1,2,4-triazol-3-yl)methanol

Hazard: Irritant

Formula: C₄H₇N₃OS
MDL Number: MFCD22192576
CAS Number: 65029-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR