4-Ethyl-1,3-oxazolidin-2-one

Supplier: Matrix Scientific

Synonyms: 4-Ethyloxazolidin-2-one

080944-500MG 080944-1G 080944-5G
102672-022EA 516.92 USD
102672-022 102672-024 102672-026
4-Ethyl-1,3-oxazolidin-2-one
4-Ethyl-1,3-oxazolidin-2-one

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD00068849
CAS Number: 16112-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR