5-Methoxybenzimidazole

Supplier: Matrix Scientific

080943-500MG 080943-1G 080943-5G
102672-016EA 516.92 USD
102672-016 102672-018 102672-020
5-Methoxybenzimidazole
5-Methoxybenzimidazole

Hazard: Irritant

Formula: C₈H₈N₂O
MDL Number: MFCD00272526
CAS Number: 4887-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR