[3-(1H-Imidazol-2-yl)propyl]amine dihydrochloride

Supplier: Matrix Scientific

080940-500MG 080940-1G 080940-5G
102671-998EA 523.08 USD
102671-998 102672-000 102672-002
[3-(1H-Imidazol-2-yl)propyl]amine dihydrochloride
[3-(1H-Imidazol-2-yl)propyl]amine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₃Cl₂N₃
MW: 198.1 g/mol
MDL Number: MFCD08668395
CAS Number: 41306-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR