3-(2-Nitroethenyl)pyridine

Supplier: Matrix Scientific

Synonyms: 3-(2-Nitrovinyl)pyridine, 2-(3-Pyridyl)-1-nitroethylene

080937-500MG 080937-1G
102671-982EA 258.46 USD
102671-982 102671-984
3-(2-Nitroethenyl)pyridine
3-(2-Nitroethenyl)pyridine

Hazard: Irritant

Formula: C₇H₆N₂O₂
MW: 150.14 g/mol
MDL Number: MFCD00191563
CAS Number: 3156-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR