2-(1H-Imidazol-1-yl)propan-1-ol

Supplier: Matrix Scientific

080933-500MG 080933-1G 080933-5G
102671-960EA 223.08 USD
102671-960 102671-962 102671-964
2-(1H-Imidazol-1-yl)propan-1-ol
2-(1H-Imidazol-1-yl)propan-1-ol

Hazard: Irritant

Formula: C₆H₁₀N₂O
MDL Number: MFCD12198461
CAS Number: 191725-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR