(3-Methoxybutyl)amine hydrochloride
Supplier: Matrix Scientific
080928-500MG
080928-1G
102671-938EA
775.38
USD
102671-938
102671-940
(3-Methoxybutyl)amine hydrochloride
(3-Methoxybutyl)amine hydrochloride
Hazard: Irritant
Formula:
C₅H₁₄ClNO |
MDL Number:
MFCD22422012 CAS Number: 1417569-76-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...