Ethyl-N-methyl-β-alaninate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-(methylamino)propanoate

080927-500MG 080927-1G
102671-934EA 516.92 USD
102671-934 102671-936
Ethyl-N-methyl-β-alaninate
Ethyl-N-methyl-beta-alaninate

Hazard: Irritant

Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
Boiling Pt: 59…61 °C (4 Torr)
Density: 1.0082/20°C
MDL Number: MFCD00672194
CAS Number: 2213-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR