1-Methyl-1,4-diazepan-5-one
Supplier: Matrix Scientific
080925-500MG
080925-1G
080925-5G
102671-924EA
516.92
USD
102671-924
102671-926
102671-928
1-Methyl-1,4-diazepan-5-one
1-Methyl-1,4-diazepan-5-one
Hazard: Irritant
Formula:
C₆H₁₂N₂O MW: 128.1700 g/mol |
MDL Number:
MFCD09027949 CAS Number: 5441-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...