5-(4-Chloro-2-nitrophenyl)-2-furoic acid

Supplier: Matrix Scientific

080921-500MG 080921-1G 080921-5G
102671-900EA 258.46 USD
102671-900 102671-902 102671-904
5-(4-Chloro-2-nitrophenyl)-2-furoic acid
5-(4-Chloro-2-nitrophenyl)-2-furoic acid

Hazard: Irritant

Formula: C₁₁H₆ClNO₅
MDL Number: MFCD00274245
CAS Number: 95611-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR