6-Methoxyquinoline-2-carboxylic acid

Supplier: Matrix Scientific

080919-500MG 080919-1G
102671-892EA 387.69 USD
102671-892 102671-894
6-Methoxyquinoline-2-carboxylic acid
6-Methoxyquinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
MDL Number: MFCD08444483
CAS Number: 75433-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR