8-Methoxyquinoline-2-carboxylic acid

Supplier: Matrix Scientific

080918-500MG 080918-1G
102671-888EA 746.15 USD
102671-888 102671-890
8-Methoxyquinoline-2-carboxylic acid
8-Methoxyquinoline-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD09030684
CAS Number: 21141-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR