4-Amino-N-methylphthalimide

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-methylisoindoline-1,3-dione

080915-500MG 080915-1G 080915-5G
102671-876EA 444.62 USD
102671-876 102671-878 102671-880
4-Amino-N-methylphthalimide
4-Amino-N-methylphthalimide

Hazard: Irritant

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
MDL Number: MFCD00012081
CAS Number: 2307-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR