N-Methyl-p-toluenesulfonamide

Supplier: Matrix Scientific

Synonyms: N,4-Dimethylbenzenesulphonamide

080913-500MG 080913-1G 080913-5G
102671-864EA 161.54 USD
102671-864 102671-866 102671-868
N-Methyl-p-toluenesulfonamide
N-Methyl-p-toluenesulfonamide

Hazard: Irritant

Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
MDL Number: MFCD00008285
CAS Number: 640-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR