[4-(Ethylsulfonyl)phenyl]amine

Supplier: Matrix Scientific

Synonyms: 4-(Ethanesulphonyl)aniline

080891-500MG 080891-1G 080891-5G
102671-786EA 387.69 USD
102671-786 102671-788 102671-790
[4-(Ethylsulfonyl)phenyl]amine
[4-(Ethylsulfonyl)phenyl]amine

Hazard: Irritant

Formula: C₈H₁₁NO₂S
MW: 185.25 g/mol
Melting Pt: 89 °C
MDL Number: MFCD01670261
CAS Number: 6334-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR