Methyl-3-oxo-3-(2-thienyl)propanoate

Supplier: Matrix Scientific

Synonyms: 3-Oxo-3-thiophen-2-yl-propionic acid methyl ester, Methyl 3-oxo-3-(thiophen-2-yl)propanoate

080877-500MG 080877-1G 080877-5G
102671-702EA 387.69 USD
102671-702 102671-704 102671-706
Methyl-3-oxo-3-(2-thienyl)propanoate
Methyl-3-oxo-3-(2-thienyl)propanoate

Hazard: Irritant

Formula: C₈H₈O₃S
MW: 184.22 g/mol
MDL Number: MFCD03424761
CAS Number: 134568-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR