Methyl-3-oxo-3-(3-thienyl)propanoate

Supplier: Matrix Scientific

080876-500MG 080876-1G 080876-5G
102671-696EA 387.69 USD
102671-696 102671-698 102671-700
Methyl-3-oxo-3-(3-thienyl)propanoate
Methyl-3-oxo-3-(3-thienyl)propanoate

Hazard: Irritant

Formula: C₈H₈O₃S
MDL Number: MFCD11974576
CAS Number: 185515-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR