2-{[Amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylic acid

Supplier: Matrix Scientific

080850-500MG 080850-1G
102671-554EA 387.69 USD
102671-554 102671-556
2-{[Amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylic acid
2-{[Amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₈N₄O₂S
MDL Number: MFCD22374974
CAS Number: 216955-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR