Methyl-2-{[amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylate

Supplier: Matrix Scientific

080839-500MG 080839-1G
102671-498EA 258.46 USD
102671-498 102671-500
Methyl-2-{[amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylate
Methyl-2-{[amino(imino)methyl]amino}-4-methyl-1,3-thiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₄O₂S
MDL Number: MFCD08131778
CAS Number: 903442-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR