N-1,3-Benzothiazol-2-ylguanidine

Supplier: Matrix Scientific

Synonyms: 1-(Benzo[d]Thiazol-2-yl)guanidine

080821-500MG 080821-1G 080821-5G
102671-398EA 258.46 USD
102671-398 102671-400 102671-402
N-1,3-Benzothiazol-2-ylguanidine
N-1,3-Benzothiazol-2-ylguanidine

Hazard: Irritant

Formula: C₈H₈N₄S
MW: 192.24 g/mol
Melting Pt: 172…173 °C
MDL Number: MFCD00022863
CAS Number: 2582-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR