N-(5-Methyl-1,3-benzoxazol-2-yl)guanidine
Supplier: Matrix Scientific
080820-500MG
080820-1G
102671-394EA
516.92
USD
102671-394
102671-396
N-(5-Methyl-1,3-benzoxazol-2-yl)guanidine
N-(5-Methyl-1,3-benzoxazol-2-yl)guanidine
Hazard: Irritant
Formula:
C₉H₁₀N₄O |
MDL Number:
MFCD01796091 CAS Number: 332898-09-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...