N-(6-Methoxy-1,3-benzothiazol-2-yl)guanidine

Supplier: Matrix Scientific

080818-500MG 080818-1G 080818-5G
102671-382EA 387.69 USD
102671-382 102671-384 102671-386
N-(6-Methoxy-1,3-benzothiazol-2-yl)guanidine
N-(6-Methoxy-1,3-benzothiazol-2-yl)guanidine

Hazard: Irritant

Formula: C₉H₁₀N₄OS
MDL Number: MFCD22374971
CAS Number: 86328-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR