N-(6-Methyl-1,3-benzothiazol-2-yl)guanidine

Supplier: Matrix Scientific

080812-500MG 080812-1G 080812-5G
102671-348EA 387.69 USD
102671-348 102671-350 102671-352
N-(6-Methyl-1,3-benzothiazol-2-yl)guanidine
N-(6-Methyl-1,3-benzothiazol-2-yl)guanidine

Hazard: Irritant

Formula: C₉H₁₀N₄S
MDL Number: MFCD02931353
CAS Number: 86328-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR