2-Mercapto-5-phenylthieno[2,3-d]pyrimidin-4(3H)-one
Supplier: Matrix Scientific
080808-500MG
080808-1G
080808-5G
102671-326EA
258.46
USD
102671-326
102671-328
102671-330
2-Mercapto-5-phenylthieno[2,3-d]pyrimidin-4(3H)-one
2-Mercapto-5-phenylthieno[2,3-d]pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₂H₈N₂OS₂ MW: 260.34 g/mol |
MDL Number:
MFCD03656342 CAS Number: 294653-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...