6-(2-Aminophenyl)-3-mercapto-1,2,4-triazin-5(4H)-one

Supplier: Matrix Scientific

080764-500MG 080764-1G
102671-140EA 193.85 USD
102671-140 102671-142
6-(2-Aminophenyl)-3-mercapto-1,2,4-triazin-5(4H)-one
6-(2-Aminophenyl)-3-mercapto-1,2,4-triazin-5(4H)-one

Hazard: Irritant

Formula: C₉H₈N₄OS
MDL Number: MFCD00636512
CAS Number: 27161-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR