6-(Chloromethyl)-N-(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080724-500MG 080724-1G 080724-5G
102670-966EA 258.46 USD
102670-966 102670-968 102670-970
6-(Chloromethyl)-N-(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(2-methoxyphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₁H₁₂ClN₅O
MDL Number: MFCD03941318
CAS Number: 578762-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR