1-(o-Tolyl)biguanide

Supplier: Matrix Scientific

080708-500MG 080708-1G 080708-5G
102670-892EA 258.46 USD
102670-892 102670-894 102670-896
1-(o-Tolyl)biguanide
1-(o-Tolyl)biguanide

Hazard: Irritant

Formula: C₉H₁₃N₅
MW: 191.2330 g/mol
MDL Number: MFCD00019731
CAS Number: 93-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR