3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
Supplier: Matrix Scientific
080701-500MG
080701-1G
102670-856EA
775.38
USD
102670-856
102670-858
3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
3-(Chloromethyl)-1-methyl-1H-pyrazole hydrochloride
Hazard: Irritant
Formula:
C₅H₈Cl₂N₂ |
MDL Number:
MFCD15143224 CAS Number: 88529-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...