(1-Methyl-1H-pyrazol-3-yl)methanol

Supplier: Matrix Scientific

080700-500MG 080700-1G 080700-5G
102670-850EA 107.69 USD
102670-850 102670-852 102670-854
(1-Methyl-1H-pyrazol-3-yl)methanol
(1-Methyl-1H-pyrazol-3-yl)methanol

Hazard: Irritant

Formula: C₅H₈N₂O
MDL Number: MFCD08690274
CAS Number: 84547-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR