3-Pyrrolidin-1-ylpropan-1-ol

Supplier: Matrix Scientific

Synonyms: 3-(1-Pyrrolidyl)-1-propanol

080698-1G 080698-5G
102670-840EA 209.23 USD
102670-840 102670-842
3-Pyrrolidin-1-ylpropan-1-ol
3-Pyrrolidin-1-ylpropan-1-ol

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD01687769
CAS Number: 19748-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR