Methyl-1-methylcyclopentanecarboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 1-methylcyclopentane-1-carboxylate

080694-500MG 080694-1G 080694-5G
102670-814EA 338.46 USD
102670-814 102670-816 102670-818
Methyl-1-methylcyclopentanecarboxylate
Methyl-1-methylcyclopentanecarboxylate

Hazard: Irritant

Formula: C₈H₁₄O₂
MDL Number: MFCD22374940
CAS Number: 4630-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR