4-(3-Chloropropyl)pyridine hydrochloride

Supplier: Matrix Scientific

080693-500MG 080693-1G 080693-5G
102670-808EA 516.92 USD
102670-808 102670-810 102670-812
4-(3-Chloropropyl)pyridine hydrochloride
4-(3-Chloropropyl)pyridine hydrochloride

Hazard: Irritant

Formula: C₈H₁₁Cl₂N
MDL Number: MFCD01691980
CAS Number: 17944-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR