5-bromoquinolin-6-amine

Supplier: Matrix Scientific

080692-500MG 080692-1G
102670-804EA 516.92 USD
102670-804 102670-806
5-bromoquinolin-6-amine
5-bromoquinolin-6-amine

Hazard: Irritant

Formula: C₉H₇BrN₂
MDL Number: MFCD00234503
CAS Number: 50358-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR