5-Amino-1,2,4-triazin-3(2H)-one

Supplier: Matrix Scientific

080688-500MG 080688-1G
102670-786EA 430.77 USD
102670-786 102670-788
5-Amino-1,2,4-triazin-3(2H)-one
5-Amino-1,2,4-triazin-3(2H)-one

Hazard: Irritant

Formula: C₃H₄N₄O
MW: 112.09 g/mol
MDL Number: MFCD00023642
CAS Number: 931-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR