2,3-Piperazinedione
Supplier: Matrix Scientific
080687-1G
080687-5G
102670-782EA
430.77
USD
102670-782
102670-784
2,3-Piperazinedione
2,3-Piperazinedione
Hazard: Irritant/Store in Freezer
Formula:
C₄H₆N₂O₂ MW: 114.1 g/mol Melting Pt: 285 °C (dec.) |
CAS Number:
13092-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...