[4-(4-Amino-3-nitrobenzyl)-2-nitrophenyl]amine

Supplier: Matrix Scientific

080685-500MG 080685-1G 080685-5G
102670-768EA 161.54 USD
102670-768 102670-770 102670-772
[4-(4-Amino-3-nitrobenzyl)-2-nitrophenyl]amine
[4-(4-Amino-3-nitrobenzyl)-2-nitrophenyl]amine

Hazard: Irritant

Formula: C₁₃H₁₂N₄O₄
MDL Number: MFCD00168261
CAS Number: 17474-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR