N-Phenyl-9H-purin-6-amine

Supplier: Matrix Scientific

080683-500MG 080683-1G 080683-5G
102670-756EA 338.46 USD
102670-756 102670-758 102670-760
N-Phenyl-9H-purin-6-amine
N-Phenyl-9H-purin-6-amine

Hazard: Irritant

Formula: C₁₁H₉N₅
MDL Number: MFCD04457455
CAS Number: 1210-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR